Search
Search
| Gene Symbol: Mas1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008552.4 | NP_032578.2 | 6 - 7 | 696 | 69 | |
| GenBank mRNA | AK015036.1 | - | 6 - 7 | 687 | 69 | |
| AK030261.1 | - | 6 - 7 | 686 | 69 | ||
| AK030265.1 | - | 6 - 7 | 697 | 69 | ||
| BC125537.1 | - | 2 - 3 | 62 | 40130052 | 69 | |
Set Membership: |
|