Search
Search

Prss52| Gene Symbol: Prss52 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028525.2 | NP_082801.2 | 3 - 4 | 597 | 80 | |
| GenBank mRNA | AB009660.1 | - | 3 - 4 | 634 | 80 | |
| AK006746.1 | - | 3 - 4 | 596 | 80 | ||
| BC049714.1 | - | 3 - 4 | 597 | 6741678 | 80 | |
Set Membership: |
|