Search
Search

Kpna3| Gene Symbol: Kpna3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008466.5 | NP_032492.1 | 8 - 9 | 709 | 115 | |
| GenBank mRNA | AF020772.1 | - | 8 - 9 | 553 | 115 | |
| AK011559.1 | - | 8 - 9 | 580 | 115 | ||
| AK049373.1 | - | 8 - 9 | 580 | 115 | ||
| AK161664.1 | - | 8 - 9 | 639 | 115 | ||
| BC026885.1 | - | 8 - 9 | 603 | 3979788 | 115 | |
Set Membership: |
|