Search
Search
| Gene Symbol: Xpo4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020506.1 | NP_065252.1 | 3 - 4 | 317 | 151 | |
| GenBank mRNA | AF145021.1 | - | 3 - 4 | 317 | 151 | |
| AK084907.1 | - | 4 - 5 | 502 | 151 | ||
| AK129430.1 | - | 3 - 4 | 312 | 151 | ||
Set Membership: |
|