Search
Search

Mmp1a| Gene Symbol: Mmp1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_032006.3 | NP_114395.1 | 4 - 5 | 673 | 68 | |
| GenBank mRNA | AJ278462.1 | - | 4 - 5 | 627 | 68 | |
| AK049552.1 | - | 4 - 5 | 674 | 68 | ||
| BC117756.2 | - | 4 - 5 | 673 | 40090601 | 68 | |
Set Membership: |
|