Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc22a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013667.2 | NP_038695.1 | 2 - 3 | 612 | 57 | |
GenBank mRNA | AJ006036.1 | - | 2 - 3 | 612 | 57 | |
AK052658.1 | - | 2 - 3 | 609 | 57 | ||
AK143892.1 | - | 2 - 3 | 611 | 57 | ||
BC015250.1 | - | 2 - 3 | 540 | 4242038 | 57 | |
BC069911.1 | - | 2 - 3 | 559 | 4974824 | 57 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |