Search
Search

Slc35f4| Gene Symbol: Slc35f4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029238.2 | NP_083514.1 | 5 - 6 | 1155 | 83 | |
| GenBank mRNA | AB041538.1 | - | 6 - 7 | 1083 | 83 | |
| AK016089.1 | - | 2 - 3 | 349 | 82 | ||
| AK034308.1 | - | 5 - 6 | 1157 | 83 | ||
| BC138981.1 | - | 5 - 6 | 1054 | 8862003 | 83 | |
| BC145720.1 | - | 5 - 6 | 1054 | 40131023 | 83 | |
Set Membership: |
|