Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01314514_m1
          See other HHIPL2 GE Assays ›
          Gene Symbol:
          Hhipl2
          Gene Name:
          hedgehog interacting protein-like 2
          Gene Aliases:
          4930507C10Rik, AW208802, Hhip3
          Chromosome Location:
          Chr.1: 183413929 - 183437053 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01314514_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 4,430.00
          Your Price
          Online offer:
          4430.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Hhipl2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK019671.1 - 8 - 9 1714 78
          BC034362.1 - 7 - 8 1459 3154701 78

          Back To Top

          More Information


          Set Membership:

          Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Mus musculus Probe spans exons

          Panther Classification:

          Molecular Function -

          protein-binding activity modulator

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P344364 BC034362.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P344364 NM_030175.4 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P344364 AK019671.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P344364 BC034362.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P344364 NM_030175.4 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P344364 AK019671.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P344364 BC034362.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P344364 NM_030175.4 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P344364 AK019671.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P344364 BC034362.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P344364 NM_030175.4 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P344364 AK019671.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P344364 BC034362.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P344364 NM_030175.4 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P344364 AK019671.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P344364 BC034362.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P344364 NM_030175.4 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P344364 AK019671.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P116097 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10598131 - -
          Affymetrix Mouse Gene 1.0 ST Array 10598134 - -
          Affymetrix Mouse Gene 1.0 ST Array 10598130 - -
          Affymetrix Mouse Gene 1.0 ST Array 10598133 - -
          Affymetrix Mouse Gene 1.0 ST Array 10598137 - -
          Affymetrix Mouse Gene 1.0 ST Array 10598132 - -
          Affymetrix Mouse Gene 1.0 ST Array 10598136 - -
          Affymetrix Mouse Gene 1.0 ST Array 10598135 - -
          Affymetrix HT MG-430 PM Array Plate 1429157_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10598126 - -
          Mouse Genome 430 2.0 Array 1429157_at - -
          Mouse Genome 430 2.0 Array 1429157_PM_at - -
          Mouse Expression Set 430 1429157_PM_at - -
          Mouse Expression Set 430 1429157_at - -
          Affymetrix HT Mouse Genome 430B Array 1429157_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1429157_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 7000026 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0007000026 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2620374 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2620374 - -
          mouse-6 v1.1 (Illumina) scl078772.9_102-S - -
          Illumina Sentrix mouse-6.v1 bead array 7000026 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl078772.9_102-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 7000026 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2620374 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2620374 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline