Search
Search

Cgrrf1| Gene Symbol: Cgrrf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026832.3 | NP_081108.2 | 2 - 3 | 277 | 79 | |
| GenBank mRNA | AK007402.1 | - | 2 - 3 | 293 | 79 | |
| AK030403.1 | - | 2 - 3 | 278 | 79 | ||
| AK030755.1 | - | 2 - 3 | 279 | 79 | ||
| AK154797.1 | - | 2 - 3 | 353 | 79 | ||
| BC114536.1 | - | 1 - 2 | 115 | 79 | ||
Set Membership: |
|