Search
Search

Dhrs4| Gene Symbol: Dhrs4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037938.2 | NP_001033027.2 | 5 - 6 | 676 | 107 | |
| GenBank mRNA | AB045132.1 | - | 5 - 6 | 588 | 107 | |
| AK002691.1 | - | 1 - 2 | 117 | 107 | ||
| AK146988.1 | - | 5 - 6 | 556 | 107 | ||
| BC003484.1 | - | 5 - 6 | 511 | 2811468 | 107 | |
| BC054361.1 | - | 5 - 6 | 637 | 5008299 | 107 | |
Set Membership: |
|