Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01317681_m1
          See other PPP3CA GE Assays ›
          Gene Symbol:
          Ppp3ca
          Gene Name:
          protein phosphatase 3, catalytic subunit, alpha isoform
          Gene Aliases:
          2900074D19Rik, CN, Caln, Calna, CnA
          Chromosome Location:
          Chr.3: 136670066 - 136937727 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01317681_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ppp3ca
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001293622.1 NP_001280551.1 5 - 6 1426 80
          NM_008913.5 NP_032939.1 5 - 6 1426 80
          GenBank mRNA AK146387.1 - 5 - 6 1370 80
          AK150393.1 - 5 - 6 785 80
          AK166287.1 - 5 - 6 1203 80
          AK189167.1 - 1 - 2 168 80
          AK211949.1 - 1 - 2 167 80
          AK213582.1 - 1 - 2 167 80
          AK219430.1 - 1 - 2 167 80
          BC138612.1 - 5 - 6 699 8861632 80
          J05479.1 - 5 - 6 723 80

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0300001382.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Protein Phosphatase Mammalian Gene Collection Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ppp3ca

          Panther Classification:

          Molecular Function -

          protein phosphatase

          Gene Ontology Categories:

          Function(s) Process(es)

          G1/S transition of mitotic cell cycle
          protein dephosphorylation
          protein import into nucleus
          calcium ion transport
          transition between fast and slow fiber
          cardiac muscle hypertrophy in response to stress
          dephosphorylation
          calcium-mediated signaling
          calcineurin-NFAT signaling cascade
          multicellular organismal response to stress
          negative regulation of chromatin binding
          cellular response to drug
          positive regulation of transcription from RNA polymerase II promoter
          negative regulation of insulin secretion
          skeletal muscle fiber development
          negative regulation of dendrite morphogenesis
          modulation of synaptic transmission
          positive regulation of sequence-specific DNA binding transcription factor activity
          positive regulation of NFAT protein import into nucleus
          response to calcium ion
          excitatory postsynaptic potential
          cellular response to glucose stimulus
          positive regulation of cardiac muscle hypertrophy in response to stress
          phosphoprotein phosphatase activity
          protein serine/threonine phosphatase activity
          calcium-dependent protein serine/threonine phosphatase activity
          protein binding
          calmodulin binding
          drug binding
          hydrolase activity
          enzyme binding
          calmodulin-dependent protein phosphatase activity
          metal ion binding
          protein heterodimerization activity
          protein dimerization activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0300011186.mm.1 NM_008913.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0300011186.mm.1 TC0300001382.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300011187.mm.1 NM_008913.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0300011187.mm.1 TC0300001382.mm.1 MTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline