Search
Search

Parp2| Gene Symbol: Parp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009632.2 | NP_033762.1 | 6 - 7 | 483 | 131 | |
| GenBank mRNA | AF072521.1 | - | 6 - 7 | 458 | 131 | |
| AJ007780.1 | - | 6 - 7 | 496 | 131 | ||
| BC052000.1 | - | 5 - 6 | 868 | 131 | ||
| BC062150.1 | - | 6 - 7 | 483 | 6333875 | 131 | |
Set Membership: |
|