Search
Search

Ngly1| Gene Symbol: Ngly1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021504.3 | NP_067479.2 | 10 - 11 | 1656 | 122 | |
| GenBank mRNA | AF250927.1 | - | 10 - 11 | 1643 | 122 | |
| AK028248.1 | - | 10 - 11 | 1657 | 122 | ||
| AK148931.1 | - | 10 - 11 | 1660 | 122 | ||
| AK169205.1 | - | 9 - 10 | 1479 | 122 | ||
| BC028961.1 | - | 10 - 11 | 1610 | 5366023 | 122 | |
Set Membership: |
|