Search
Search
| Gene Symbol: Adh4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011996.2 | NP_036126.2 | 3 - 4 | 298 | 79 | |
| GenBank mRNA | AJ245750.1 | - | 3 - 4 | 286 | 79 | |
| AK132994.1 | - | 3 - 4 | 298 | 79 | ||
| BC100729.1 | - | 3 - 4 | 286 | 40045447 | 79 | |
| BC100730.1 | - | 3 - 4 | 285 | 40045448 | 79 | |
| BC100736.1 | - | 3 - 4 | 286 | 40045445 | 79 | |
Set Membership: |
|