Search
Search

Larp7| Gene Symbol: Larp7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138593.2 | NP_613059.2 | 3 - 4 | 457 | 102 | |
| GenBank mRNA | AK012772.1 | - | 3 - 4 | 447 | 102 | |
| AK015041.1 | - | 3 - 4 | 432 | 102 | ||
| AK082815.1 | - | 3 - 4 | 456 | 102 | ||
| AK162147.1 | - | 3 - 4 | 374 | 102 | ||
| BC020127.1 | - | 2 - 3 | 308 | 102 | ||
| BC023165.1 | - | 3 - 4 | 411 | 102 | ||
Set Membership: |
|