Search
Search

Myoz2| Gene Symbol: Myoz2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021503.2 | NP_067478.1 | 2 - 3 | 262 | 117 | |
| GenBank mRNA | AJ252148.1 | - | 2 - 3 | 224 | 117 | |
| AK003634.1 | - | 2 - 3 | 214 | 115 | ||
| AK085927.1 | - | 2 - 3 | 227 | 117 | ||
| AY013296.1 | - | 2 - 3 | 238 | 117 | ||
| BC024360.1 | - | 2 - 3 | 224 | 5012436 | 117 | |
Set Membership: |
|