Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01321738_m1
          See other TGFB2 GE Assays ›
          Gene Symbol:
          Tgfb2
          Gene Name:
          transforming growth factor, beta 2
          Gene Aliases:
          BB105277, Tgf-beta2, Tgfb-2
          Chromosome Location:
          Chr.1: 186622787 - 186709033 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01321738_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tgfb2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_009367.3 NP_033393.2 5 - 6 2150 61
          GenBank mRNA AK029306.1 - 3 - 4 362 61
          AK159690.1 - 5 - 6 714 61
          BC011055.1 - 3 - 4 513 3585774 61
          BC011170.1 - 5 - 6 1058 4035366 61
          X57413.1 - 5 - 6 2149 61

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0100003754.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tgfb2

          Panther Classification:

          Molecular Function -

          growth factor

          Gene Ontology Categories:

          Function(s) Process(es)

          cell morphogenesis
          skeletal system development
          cartilage condensation
          blood vessel development
          eye development
          response to hypoxia
          epithelial to mesenchymal transition
          hair follicle development
          blood vessel remodeling
          heart morphogenesis
          outflow tract morphogenesis
          heart valve morphogenesis
          cardiac left ventricle morphogenesis
          cardiac right ventricle morphogenesis
          protein phosphorylation
          cell cycle arrest
          transforming growth factor beta receptor signaling pathway
          SMAD protein import into nucleus
          axon guidance
          heart development
          cell death
          positive regulation of cell proliferation
          negative regulation of cell proliferation
          glial cell migration
          cardioblast differentiation
          positive regulation of gene expression
          positive regulation of epithelial cell migration
          negative regulation of alkaline phosphatase activity
          positive regulation of epithelial to mesenchymal transition
          positive regulation of pathway-restricted SMAD protein phosphorylation
          negative regulation of macrophage cytokine production
          positive regulation of phosphatidylinositol 3-kinase signaling
          cell growth
          cell migration
          signal transduction by protein phosphorylation
          hemopoiesis
          extracellular matrix organization
          collagen fibril organization
          positive regulation of cell growth
          negative regulation of cell growth
          positive regulation of cell migration
          hair follicle morphogenesis
          activation of protein kinase activity
          response to progesterone
          positive regulation of stress-activated MAPK cascade
          regulation of transforming growth factor beta2 production
          positive regulation of cell adhesion mediated by integrin
          intracellular signal transduction
          growth
          wound healing
          regulation of cell proliferation
          dopamine biosynthetic process
          response to drug
          catagen
          regulation of apoptotic process
          positive regulation of apoptotic process
          negative regulation of apoptotic process
          regulation of MAPK cascade
          positive regulation of neuron apoptotic process
          response to estrogen
          cell-cell junction organization
          negative regulation of keratinocyte differentiation
          positive regulation of integrin biosynthetic process
          positive regulation of cell cycle
          positive regulation of heart contraction
          somatic stem cell division
          neuron fate commitment
          neuron development
          embryonic neurocranium morphogenesis
          negative regulation of epithelial cell proliferation
          positive regulation of protein secretion
          negative regulation of immune response
          negative regulation of release of sequestered calcium ion into cytosol
          positive regulation of cell division
          positive regulation of catagen
          positive regulation of cardioblast differentiation
          cardiac muscle cell proliferation
          cardiac epithelial to mesenchymal transition
          pathway-restricted SMAD protein phosphorylation
          SMAD protein signal transduction
          ventricular septum morphogenesis
          negative regulation of cartilage development
          positive regulation of activation-induced cell death of T cells
          extrinsic apoptotic signaling pathway
          regulation of extracellular matrix organization
          positive regulation of extrinsic apoptotic signaling pathway in absence of ligand
          beta-amyloid binding
          receptor signaling protein serine/threonine kinase activity
          receptor binding
          type II transforming growth factor beta receptor binding
          cytokine activity
          transforming growth factor beta receptor binding
          protein binding
          growth factor activity
          type III transforming growth factor beta receptor binding
          protein homodimerization activity
          protein heterodimerization activity
          protein N-terminus binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0100030757.mm.1 NM_009367.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0100030757.mm.1 TC0100003754.mm.1 MTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline