Search
Search
| Gene Symbol: Gpc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008150.2 | NP_032176.2 | 1 - 2 | 309 | 104 | |
| GenBank mRNA | BC006622.1 | - | 1 - 2 | 309 | 3967797 | 104 |
| CJ187434.1 | - | 1 - 2 | 263 | 104 | ||
| X83577.1 | - | 1 - 2 | 630 | 104 | ||
Set Membership: |
|