Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01322668_s1
          See other CCDC89 GE Assays ›
          Gene Symbol:
          Ccdc89
          Gene Name:
          coiled-coil domain containing 89
          Gene Aliases:
          1700019B01Rik, AU022100, BOIP
          Chromosome Location:
          Chr.7: 90426312 - 90428664 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01322668_s1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ccdc89
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_027298.1 NP_081574.1 1 1451 128
          GenBank mRNA BC060150.1 - 1 1185 128

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P169445 BC060150.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P169445 AK006105.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P169445 NM_027298.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P169445 BC060150.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P169445 AK006105.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P169445 NM_027298.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 465168 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 113077 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P116965 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P169445 BC060150.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P169445 AK006105.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P169445 NM_027298.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P169445 BC060150.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P169445 AK006105.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P169445 NM_027298.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P116965 - -
          Whole Mouse Genome G4122A A_51_P169445 BC060150.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P169445 AK006105.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P169445 NM_027298.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P169445 BC060150.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P169445 AK006105.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P169445 NM_027298.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P116965 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25313531 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25313537 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25313552 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25313576 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25313553 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05958762 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25313548 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25313554 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05958752 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25313541 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25313578 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05958764 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25313572 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06293483 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25313577 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25313550 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25313533 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25313535 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25313528 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25313529 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25313555 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25313545 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25313573 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25313544 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10554894 - -
          Affymetrix HT MG-430 PM Array Plate 1429635_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1453211_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10554893 - -
          Mouse Genome 430 2.0 Array 1453211_PM_at - -
          Mouse Genome 430 2.0 Array 1429635_PM_at - -
          Mouse Genome 430 2.0 Array 1429635_at - -
          Mouse Genome 430 2.0 Array 1453211_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 70054_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 70054_PM_at - -
          Mouse Expression Set 430 1429635_at - -
          Mouse Expression Set 430 1429635_PM_at - -
          Mouse Expression Set 430 1453211_at - -
          Mouse Expression Set 430 1453211_PM_at - -
          Murine Genome U74v2 Set 166386_at - -
          Murine Genome U74v2 Set 166386_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1429635_at - -
          Affymetrix HT Mouse Genome 430B Array 1453211_at - -
          Affymetrix HT Mouse Genome 430B Array 1429635_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1453211_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 727922 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2736703 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2711321 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2736703 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline