Search
Search

Usp26| Gene Symbol: Usp26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031388.2 | NP_113565.2 | 3 - 4 | 329 | 62 | |
| GenBank mRNA | AK082829.1 | - | 3 - 4 | 256 | 62 | |
| AK133417.1 | - | 3 - 4 | 333 | 62 | ||
Set Membership: |
|