Search
Search

Pcf11| Gene Symbol: Pcf11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029078.3 | NP_083354.3 | 7 - 8 | 2384 | 94 | |
| GenBank mRNA | AK173035.1 | - | 7 - 8 | 2356 | 94 | |
| BC150781.1 | - | 7 - 8 | 2220 | 9087692 | 94 | |
| EF537860.1 | - | 4 - 5 | 1488 | 94 | ||
| EF537861.1 | - | 4 - 5 | 1488 | 94 | ||
Set Membership: |
|