Search
Search

Naa16| Gene Symbol: Naa16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025832.2 | NP_080108.1 | 4 - 5 | 462 | 67 | |
| GenBank mRNA | AK005056.1 | - | 4 - 5 | 472 | 67 | |
| AK007755.1 | - | 4 - 5 | 498 | 67 | ||
| AK028432.1 | - | 4 - 5 | 572 | 67 | ||
| AK152498.1 | - | 4 - 5 | 472 | 67 | ||
| BC052445.1 | - | 4 - 5 | 462 | 6412745 | 67 | |
Set Membership: |
|