Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01325122_m1
          See other AKR1C19 GE Assays ›
          Gene Symbol:
          Akr1c19
          Gene Name:
          aldo-keto reductase family 1, member C19
          Gene Aliases:
          1810010N06Rik
          Chromosome Location:
          Chr.13: 4233504 - 4248361 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01325122_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Akr1c19
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001013785.3 NP_001013807.2 8 - 9 959 139
          GenBank mRNA BC087964.1 - 8 - 9 959 6767816 139
          BG073853.2 - 1 - 2 139

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1300000020.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Akr1c19

          Panther Classification:

          Molecular Function -

          oxidoreductase reductase metabolite interconversion enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2007919 NM_001013785.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2007919 BC087964.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 590907 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 475822 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 346254 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 457270 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 434046 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 242554 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 145162 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 286177 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 68893 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 142612 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2007919 NM_001013785.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2007919 BC087964.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P180213 NM_001013785.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P180213 BC087964.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P111335 NM_001013785.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P111335 BC087964.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28791781 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28791798 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28791750 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28791783 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28791742 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28791737 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28791794 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28791758 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28791797 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28791784 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28791755 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28791743 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28791773 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28791792 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28791769 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28791752 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28791745 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28791757 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28791785 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28791741 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28791759 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28791767 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28791775 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28791778 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28791780 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28791788 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28791763 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28791762 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28791748 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28791787 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28791764 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28791779 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28791776 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28791747 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28791751 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28791790 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28791796 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28791736 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28791744 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28791739 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28791774 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28791795 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07772044 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07772024 - -
          Whole Mouse Genome G4122A A_51_P180213 NM_001013785.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P180213 BC087964.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P111335 NM_001013785.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P111335 BC087964.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P07772024 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11103854 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11103848 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11103855 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11103850 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11103851 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11103857 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28791886 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28791752 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28791841 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28791864 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28791785 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28791836 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28791818 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28791778 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28791763 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28791767 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28791830 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28791806 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28791735 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28791860 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28791865 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28791794 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28791773 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28791770 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28791839 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28791743 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28791775 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28791809 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28791871 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28791742 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28791737 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28791796 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28791724 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28791781 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28791829 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28791759 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28791744 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28791748 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28791758 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28791776 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28791872 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28791798 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28791850 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28791852 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28791757 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28791844 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28791848 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28791890 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28791831 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28791804 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28791821 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28791811 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28791842 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28791866 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28791805 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28791731 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28791812 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28791802 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28791834 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28791874 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28791755 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28791892 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28791764 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28791747 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28791762 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28791783 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28791835 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28791896 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28791792 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28791838 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28791824 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28791800 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28791862 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28791739 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28791751 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28791858 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28791888 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28791867 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28791868 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28791825 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28791816 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28791824 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10628271 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28791783 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28791825 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28791816 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11103848 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28791796 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28791751 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28791750 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28791767 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28791741 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28791808 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28791787 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11103857 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28791752 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28791799 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28791774 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28791758 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28791778 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28791802 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28791818 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28791731 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28791728 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28791763 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28791781 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28791795 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28791821 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28791822 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28791804 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10527626 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28791806 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28791748 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10690001 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10476355 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28791790 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28791797 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28791773 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28791812 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28791798 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28791805 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28791794 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28791747 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28791809 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28791759 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28791726 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28791792 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28791776 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28791814 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28791742 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28791737 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10622117 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28791779 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28791785 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10680375 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28791755 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28791811 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11103851 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28791801 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28791729 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28791735 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10690000 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28791800 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28791826 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28791764 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28791727 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28791757 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10403318 - -
          Affymetrix Mouse Gene 1.0 ST Array 10403313 - -
          Affymetrix Mouse Gene 1.0 ST Array 10403321 - -
          Affymetrix Mouse Gene 1.0 ST Array 10403314 - -
          Affymetrix Mouse Gene 1.0 ST Array 10403320 - -
          Affymetrix Mouse Gene 1.0 ST Array 10403315 - -
          Affymetrix Mouse Gene 1.0 ST Array 10403317 - -
          Affymetrix Mouse Gene 1.0 ST Array 10403316 - -
          Affymetrix Mouse Gene 1.0 ST Array 10403319 - -
          Affymetrix HT MG-430 PM Array Plate 1455454_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10403312 - -
          Mouse Genome 430 2.0 Array 1455454_at - -
          Mouse Genome 430 2.0 Array 1455454_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 432720_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 432720_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1300000161.mm.1 NM_001013785.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300000161.mm.1 NM_001013785.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1300000161.mm.1 TC1300000020.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300000162.mm.1 NM_001013785.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300000162.mm.1 NM_001013785.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1300000162.mm.1 TC1300000020.mm.1 MTA TC
          Mouse Expression Set 430 1455454_PM_at - -
          Mouse Expression Set 430 1455454_at - -
          Affymetrix HT Mouse Genome 430B Array 1455454_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1455454_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 727935 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_3160292 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_3160292 - -
          mouse-6 v1.1 (Illumina) GI_62000659-S - -
          MouseRef-8 v2 Expression BeadChip ILMN_3160292 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3160292 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline