Search
Search
| Gene Symbol: Ctc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013256.2 | NP_001013274.2 | 6 - 7 | 1293 | 63 | |
| NM_001143790.1 | NP_001137262.1 | 6 - 7 | 1293 | 63 | ||
| NM_001281464.1 | NP_001268393.1 | 6 - 7 | 1164 | 63 | ||
| NM_001281465.1 | NP_001268394.1 | 6 - 7 | 1277 | 63 | ||
| GenBank mRNA | AK038651.1 | - | 6 - 7 | 1080 | 63 | |
| AK044055.1 | - | 6 - 7 | 1072 | 63 | ||
| AK145919.1 | - | 6 - 7 | 1108 | 63 | ||
| AK165906.1 | - | 6 - 7 | 1093 | 63 | ||
| BC060629.1 | - | 2 - 3 | 343 | 6847619 | 63 | |
| BC066078.1 | - | 6 - 7 | 1108 | 63 | ||
| BC072625.1 | - | 6 - 7 | 1121 | 63 | ||
| BC147495.1 | - | 6 - 7 | 1091 | 9056325 | 63 | |
| BC147497.1 | - | 6 - 7 | 1091 | 9056354 | 63 | |
Set Membership: |
|