Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01325954_g1
          See other EIF5 GE Assays ›
          Gene Symbol:
          Eif5
          Gene Name:
          eukaryotic translation initiation factor 5
          Gene Aliases:
          2810011H21Rik, D12Ertd549e
          Chromosome Location:
          Chr.12: 111538101 - 111546753 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01325954_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Eif5
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_173363.5 NP_775539.1 1 - 2 141 93
          NM_178041.2 NP_829887.1 1 - 2 141 93
          GenBank mRNA AI391022.1 - 1 - 2 141 93
          AK161554.1 - 1 - 2 73 93
          BC039275.1 - 1 - 2 37 4984912 93

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1200001239.mm.1

          Additional Information:

          This assay is in the 5'UTR region of the AK161554.1,BC039275.1,NM_173363.5,NM_178041.2 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Eif5

          Panther Classification:

          Molecular Function -

          translation initiation factor

          Gene Ontology Categories:

          Function(s) Process(es)

          translation
          translational initiation
          regulation of translational initiation
          nucleotide binding
          translation initiation factor activity
          GTP binding
          poly(A) RNA binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1200008781.mm.1 NM_173363.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200008781.mm.1 NM_173363.5 Refseq
          Affymetrix Mouse Clariom S Array PSR1200008781.mm.1 NM_178041.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200008781.mm.1 NM_178041.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1200008781.mm.1 TC1200001239.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200008789.mm.1 NM_173363.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200008789.mm.1 NM_173363.5 Refseq
          Affymetrix Mouse Clariom S Array PSR1200008789.mm.1 NM_178041.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200008789.mm.1 NM_178041.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1200008789.mm.1 TC1200001239.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200008795.mm.1 NM_173363.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200008795.mm.1 NM_173363.5 Refseq
          Affymetrix Mouse Clariom S Array PSR1200008795.mm.1 NM_178041.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200008795.mm.1 NM_178041.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1200008795.mm.1 TC1200001239.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200008805.mm.1 NM_173363.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200008805.mm.1 NM_173363.5 Refseq
          Affymetrix Mouse Clariom S Array PSR1200008805.mm.1 NM_178041.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200008805.mm.1 NM_178041.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1200008805.mm.1 TC1200001239.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200008782.mm.1 NM_173363.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200008782.mm.1 NM_173363.5 Refseq
          Affymetrix Mouse Clariom S Array PSR1200008782.mm.1 NM_178041.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200008782.mm.1 NM_178041.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1200008782.mm.1 TC1200001239.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200008787.mm.1 NM_173363.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200008787.mm.1 NM_173363.5 Refseq
          Affymetrix Mouse Clariom S Array PSR1200008787.mm.1 TC1200001239.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200008802.mm.1 NM_173363.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200008802.mm.1 NM_173363.5 Refseq
          Affymetrix Mouse Clariom S Array PSR1200008802.mm.1 NM_178041.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200008802.mm.1 NM_178041.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1200008802.mm.1 TC1200001239.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200008803.mm.1 NM_173363.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200008803.mm.1 NM_173363.5 Refseq
          Affymetrix Mouse Clariom S Array PSR1200008803.mm.1 NM_178041.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200008803.mm.1 NM_178041.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1200008803.mm.1 TC1200001239.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200008783.mm.1 NM_173363.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200008783.mm.1 NM_173363.5 Refseq
          Affymetrix Mouse Clariom S Array PSR1200008783.mm.1 NM_178041.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200008783.mm.1 NM_178041.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1200008783.mm.1 TC1200001239.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200008804.mm.1 NM_173363.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200008804.mm.1 NM_173363.5 Refseq
          Affymetrix Mouse Clariom S Array PSR1200008804.mm.1 NM_178041.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200008804.mm.1 NM_178041.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1200008804.mm.1 TC1200001239.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200008788.mm.1 NM_173363.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200008788.mm.1 NM_173363.5 Refseq
          Affymetrix Mouse Clariom S Array PSR1200008788.mm.1 TC1200001239.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200008790.mm.1 NM_173363.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200008790.mm.1 NM_173363.5 Refseq
          Affymetrix Mouse Clariom S Array PSR1200008790.mm.1 NM_178041.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200008790.mm.1 NM_178041.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1200008790.mm.1 TC1200001239.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200008793.mm.1 NM_173363.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200008793.mm.1 NM_173363.5 Refseq
          Affymetrix Mouse Clariom S Array PSR1200008793.mm.1 NM_178041.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200008793.mm.1 NM_178041.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1200008793.mm.1 TC1200001239.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200008806.mm.1 NM_173363.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200008806.mm.1 NM_173363.5 Refseq
          Affymetrix Mouse Clariom S Array PSR1200008806.mm.1 NM_178041.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200008806.mm.1 NM_178041.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1200008806.mm.1 TC1200001239.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200008807.mm.1 NM_173363.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200008807.mm.1 NM_173363.5 Refseq
          Affymetrix Mouse Clariom S Array PSR1200008807.mm.1 NM_178041.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200008807.mm.1 NM_178041.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1200008807.mm.1 TC1200001239.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 596226 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline