Search
Search

Usp20| Gene Symbol: Usp20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028846.5 | NP_083122.1 | 3 - 4 | 320 | 69 | |
| GenBank mRNA | AF449715.1 | - | 3 - 4 | 271 | 69 | |
| AK029890.1 | - | 217 | 69 | |||
| AK042612.1 | - | 3 - 4 | 237 | 69 | ||
| AK047242.1 | - | 3 - 4 | 241 | 69 | ||
| AK054279.1 | - | 3 - 4 | 263 | 69 | ||
| AK087412.1 | - | 3 - 4 | 231 | 69 | ||
| AK163663.1 | - | 3 - 4 | 230 | 69 | ||
| AK173083.1 | - | 3 - 4 | 222 | 69 | ||
| BC079674.1 | - | 209 | 30531994 | 69 | ||
| BY251374.1 | - | 3 - 4 | 321 | 69 | ||
Set Membership: |
|