Search
Search

Uimc1| Gene Symbol: Uimc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011307.2 | NP_035437.1 | 5 - 6 | 570 | 75 | |
| NR_121592.1 | 5 - 6 | 575 | 75 | |||
| GenBank mRNA | AK052704.1 | - | 5 - 6 | 571 | 75 | |
| AK083342.1 | - | 5 - 6 | 511 | 75 | ||
| BC037092.1 | - | 5 - 6 | 531 | 75 | ||
| BC040808.1 | - | 5 - 6 | 526 | 4038778 | 75 | |
| BC051659.1 | - | 5 - 6 | 493 | 75 | ||
| BC066220.1 | - | 5 - 6 | 493 | 75 | ||
Set Membership: |
|