Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01327285_g1
          See other BTBD6 GE Assays ›
          Gene Symbol:
          Btbd6
          Gene Name:
          BTB (POZ) domain containing 6
          Gene Aliases:
          -
          Chromosome Location:
          Chr.12: 112974998 - 112979493 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01327285_g1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Btbd6
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001145900.1 NP_001139372.1 4 - 5 549 59
          NM_201646.2 NP_964008.2 3 - 4 631 59
          GenBank mRNA AK146991.1 - 3 - 4 463 59
          BC031195.1 - 3 - 4 631 5059510 59

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1200001275.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Btbd6

          Panther Classification:

          Molecular Function -

          ubiquitin-protein ligase

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P635271 BC031195.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P635271 BC031195.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P147388 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P635271 BC031195.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P635271 BC031195.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P635271 BC031195.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P635271 BC031195.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P130999 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10167045 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10167044 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10167047 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10398931 - -
          Affymetrix Mouse Gene 1.0 ST Array 10398932 - -
          Affymetrix Mouse Gene 1.0 ST Array 10398933 - -
          Affymetrix Mouse Gene 1.0 ST Array 10398930 - -
          Affymetrix HT MG-430 PM Array Plate 1435153_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10398929 - -
          Mouse Genome 430 2.0 Array 1435153_PM_at - -
          Mouse Genome 430 2.0 Array 1435153_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 399566_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 399566_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1200009182.mm.1 NM_001145900.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200009182.mm.1 NM_001145900.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200009182.mm.1 NM_201646.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200009182.mm.1 NM_201646.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1200009182.mm.1 TC1200001275.mm.1 MTA TC
          Mouse Expression Set 430 1435153_at - -
          Mouse Expression Set 430 1435153_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1435153_at - -
          Affymetrix HT Mouse Genome 430B Array 1435153_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 407268 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 110088 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000110088 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2774822 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2774822 - -
          mouse-6 v1.1 (Illumina) scl0399566.4_97-S - -
          Illumina Sentrix mouse-6.v1 bead array 110088 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0399566.4_97-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 110088 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2839682 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2774822 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2839682 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl0399566.4_97-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline