| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0200035299.mm.1 |
NM_001083810.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200035299.mm.1 |
NM_001083810.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200035299.mm.1 |
NM_001110849.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200035299.mm.1 |
NM_001110849.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200035299.mm.1 |
NM_175181.5.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200035299.mm.1 |
NM_175181.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200035299.mm.1 |
TC0200004204.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200035301.mm.1 |
NM_001083810.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200035301.mm.1 |
NM_001083810.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200035301.mm.1 |
NM_001110849.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200035301.mm.1 |
NM_001110849.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200035301.mm.1 |
NM_175181.5.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200035301.mm.1 |
NM_175181.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200035301.mm.1 |
TC0200004204.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200035302.mm.1 |
NM_001083810.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200035302.mm.1 |
NM_001083810.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200035302.mm.1 |
TC0200004204.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200035305.mm.1 |
NM_001083810.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200035305.mm.1 |
NM_001083810.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200035305.mm.1 |
NM_175181.5.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200035305.mm.1 |
NM_175181.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200035305.mm.1 |
TC0200004204.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200035311.mm.1 |
NM_001110849.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200035311.mm.1 |
NM_001110849.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200035311.mm.1 |
TC0200004204.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200035293.mm.1 |
NM_001083810.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200035293.mm.1 |
NM_001083810.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200035293.mm.1 |
NM_001110849.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200035293.mm.1 |
NM_001110849.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200035293.mm.1 |
NM_175181.5.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200035293.mm.1 |
NM_175181.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200035293.mm.1 |
TC0200004204.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200035304.mm.1 |
NM_001083810.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200035304.mm.1 |
NM_001083810.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200035304.mm.1 |
NM_175181.5.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200035304.mm.1 |
NM_175181.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200035304.mm.1 |
TC0200004204.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200035303.mm.1 |
NM_001083810.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200035303.mm.1 |
NM_001083810.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200035303.mm.1 |
NM_175181.5.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200035303.mm.1 |
NM_175181.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200035303.mm.1 |
TC0200004204.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200035296.mm.1 |
NM_001083810.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200035296.mm.1 |
NM_001083810.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200035296.mm.1 |
NM_001110849.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200035296.mm.1 |
NM_001110849.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200035296.mm.1 |
NM_175181.5.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200035296.mm.1 |
NM_175181.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200035296.mm.1 |
TC0200004204.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200035298.mm.1 |
NM_001083810.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200035298.mm.1 |
NM_001083810.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200035298.mm.1 |
NM_001110849.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200035298.mm.1 |
NM_001110849.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200035298.mm.1 |
NM_175181.5.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200035298.mm.1 |
NM_175181.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200035298.mm.1 |
TC0200004204.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200035290.mm.1 |
NM_001083810.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200035290.mm.1 |
NM_001083810.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200035290.mm.1 |
NM_001110849.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200035290.mm.1 |
NM_001110849.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200035290.mm.1 |
NM_175181.5.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200035290.mm.1 |
NM_175181.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200035290.mm.1 |
TC0200004204.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200035300.mm.1 |
NM_001083810.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200035300.mm.1 |
NM_001083810.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200035300.mm.1 |
NM_001110849.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200035300.mm.1 |
NM_001110849.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200035300.mm.1 |
NM_175181.5.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200035300.mm.1 |
NM_175181.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200035300.mm.1 |
TC0200004204.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200035292.mm.1 |
NM_001083810.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200035292.mm.1 |
NM_001083810.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200035292.mm.1 |
NM_001110849.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200035292.mm.1 |
NM_001110849.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200035292.mm.1 |
NM_175181.5.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200035292.mm.1 |
NM_175181.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200035292.mm.1 |
TC0200004204.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200035312.mm.1 |
NM_001110849.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200035312.mm.1 |
NM_001110849.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200035312.mm.1 |
TC0200004204.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200035291.mm.1 |
NM_001083810.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200035291.mm.1 |
NM_001083810.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200035291.mm.1 |
NM_001110849.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200035291.mm.1 |
NM_001110849.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200035291.mm.1 |
NM_175181.5.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200035291.mm.1 |
NM_175181.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200035291.mm.1 |
TC0200004204.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200035295.mm.1 |
NM_001083810.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200035295.mm.1 |
NM_001083810.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200035295.mm.1 |
NM_001110849.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200035295.mm.1 |
NM_001110849.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200035295.mm.1 |
NM_175181.5.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200035295.mm.1 |
NM_175181.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200035295.mm.1 |
TC0200004204.mm.1 |
MTA TC |