Search
Search

Rabl6| Gene Symbol: Rabl6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024616.1 | NP_001019787.2 | 5 - 6 | 660 | 98 | |
| GenBank mRNA | AK045520.1 | - | 5 - 6 | 682 | 98 | |
| AK081440.1 | - | 5 - 6 | 670 | 98 | ||
| AK143678.1 | - | 5 - 6 | 715 | 98 | ||
| AK157020.1 | - | 5 - 6 | 696 | 98 | ||
| AK171643.1 | - | 5 - 6 | 671 | 98 | ||
| BC020111.1 | - | 5 - 6 | 668 | 98 | ||
| BC021476.1 | - | 5 - 6 | 675 | 98 | ||
| BC023692.1 | - | 5 - 6 | 688 | 98 | ||
| BC051138.1 | - | 5 - 6 | 674 | 98 | ||
| BC058585.1 | - | 5 - 6 | 673 | 98 | ||
| BC085507.1 | - | 5 - 6 | 660 | 5716088 | 98 | |
Set Membership: |
|