Search
Search
| Gene Symbol: Epn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252454.1 | NP_001239383.1 | 5 - 6 | 950 | 81 | |
| NM_010147.4 | NP_034277.1 | 6 - 7 | 1121 | 81 | ||
| GenBank mRNA | AK155033.1 | - | 6 - 7 | 1116 | 81 | |
| BC046962.1 | - | 4 - 5 | 756 | 6391968 | 81 | |
| BC067206.1 | - | 5 - 6 | 892 | 6514327 | 81 | |
| BC099682.1 | - | 6 - 7 | 1051 | 6529672 | 81 | |
Set Membership: |
|