Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01328503_m1
          See other RFPL4 GE Assays ›
          Gene Symbol:
          Rfpl4
          Gene Name:
          ret finger protein-like 4
          Gene Aliases:
          D7Ertd486e, Rfpl4a
          Chromosome Location:
          Chr.7: 5109787 - 5116911 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01328503_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rfpl4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001145013.1 NP_001138485.1 2 - 3 332 90
          NM_138954.3 NP_620404.1 2 - 3 564 90
          GenBank mRNA AK136022.1 - 2 - 3 333 90
          AK139569.1 - 2 - 3 566 90
          AY070253.1 - 2 - 3 554 90
          BC069834.1 - 2 - 3 318 30474534 90

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700002185.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rfpl4

          Gene Ontology Categories:

          Process(es)

          protein binding
          zinc ion binding
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P400976 AY070253.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P400976 BC069834.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P400976 NM_001145013.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P400976 NM_138954.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P400976 AK136022.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P400976 AY070253.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P400976 BC069834.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P400976 NM_001145013.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P400976 NM_138954.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P400976 AK136022.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 683191 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 361728 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 439863 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 547345 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 59667 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P127774 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P400976 AY070253.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P400976 BC069834.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P400976 NM_001145013.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P400976 NM_138954.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P400976 AK136022.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P400976 AY070253.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P400976 BC069834.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P400976 NM_001145013.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P400976 NM_138954.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P400976 AK136022.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24952227 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24952228 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24952233 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24952241 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24952238 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24952234 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24952243 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24952230 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24952250 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24952221 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24952226 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24952245 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24952248 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24952240 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24952229 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24952236 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24952223 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24952249 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24952247 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01476440 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P127774 - -
          Whole Mouse Genome G4122A A_51_P400976 AY070253.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P400976 BC069834.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P400976 NM_001145013.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P400976 NM_138954.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P400976 AK136022.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P400976 AY070253.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P400976 BC069834.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P400976 NM_001145013.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P400976 NM_138954.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P400976 AK136022.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P115626 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P127774 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06183234 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06183232 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06638543 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06183233 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06183229 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06183235 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24952227 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24952222 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24952201 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24952243 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24952189 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24952210 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24952220 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24952247 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24952197 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24952221 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24952195 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24952234 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24952262 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24952229 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24952240 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24952208 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24952250 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24952199 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24952172 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24952206 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24952226 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24952213 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24952182 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24952204 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24952263 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24952179 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24952248 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24952214 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24952184 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24952216 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24952249 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24952192 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24952174 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24952178 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24952219 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24952209 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24952233 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24952200 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24952223 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24952230 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24952217 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24952245 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24952193 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24952238 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24952201 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24952217 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24952216 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24952209 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24952228 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06255142 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24952234 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24952192 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06129693 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24952250 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06135000 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06638543 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24952222 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24952199 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24952230 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06390007 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05982776 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24952204 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24952203 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06146335 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24952219 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24952220 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24952221 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24952200 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24952229 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06255138 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06255141 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24952240 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06540831 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06134995 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24952227 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24952193 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24952213 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24952233 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06255139 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24952206 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24952208 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06540826 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10559717 - -
          Affymetrix Mouse Gene 1.0 ST Array 10559719 - -
          Affymetrix Mouse Gene 1.0 ST Array 10559718 - -
          Affymetrix HT MG-430 PM Array Plate 1416650_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10559716 - -
          Mouse Genome 430 2.0 Array 1416650_PM_at - -
          Mouse Genome 430 2.0 Array 1416650_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 192658_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 192658_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0700019978.mm.1 NM_001145013.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700019978.mm.1 NM_001145013.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700019978.mm.1 NM_138954.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700019978.mm.1 NM_138954.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0700019978.mm.1 TC0700002185.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700019979.mm.1 NM_001145013.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700019979.mm.1 NM_001145013.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700019979.mm.1 NM_138954.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700019979.mm.1 NM_138954.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0700019979.mm.1 TC0700002185.mm.1 MTA TC
          Mouse Expression Set 430 1416650_PM_at - -
          Mouse Expression Set 430 1416650_at - -
          Murine Genome U74v2 Set 93449_at - -
          Murine Genome U74v2 Set 93449_PM_at - -
          Mouse Genome 430A 2.0 Array 1416650_PM_at - -
          Mouse Genome 430A 2.0 Array 1416650_at - -
          Affymetrix HT Mouse Genome 430A Array 1416650_at - -
          Affymetrix HT Mouse Genome 430A Array 1416650_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 762386 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 4760280 - -
          Illumina mouse-6 v1.0 expression beadchip 4760280 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004760280 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2605489 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2605489 - -
          mouse-6 v1.1 (Illumina) scl31784.3_265-S - -
          Illumina Sentrix mouse-6.v1 bead array 4760280 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl31784.3_265-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4760280 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2872334 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2605489 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2872334 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2605489 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline