Search
Search

Stk31| Gene Symbol: Stk31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029916.2 | NP_084192.2 | 3 - 4 | 180 | 152 | |
| GenBank mRNA | AF285580.1 | - | 3 - 4 | 180 | 152 | |
| BC137989.1 | - | 3 - 4 | 177 | 8861008 | 152 | |
Set Membership: |
|