| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0600004462.mm.1 |
NM_029353.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600004462.mm.1 |
NM_029353.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600004462.mm.1 |
TC0600000578.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600004463.mm.1 |
NM_029353.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600004463.mm.1 |
NM_029353.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600004463.mm.1 |
TC0600000578.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600004464.mm.1 |
NM_029353.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600004464.mm.1 |
NM_029353.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600004464.mm.1 |
TC0600000578.mm.1 |
MTA TC |