Search
Search
| Gene Symbol: Gas7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109657.3 | NP_001103127.1 | 3 - 4 | 520 | 110 | |
| NM_001277079.1 | NP_001264008.1 | 3 - 4 | 349 | 110 | ||
| NM_008088.5 | NP_032114.2 | 3 - 4 | 326 | 110 | ||
| GenBank mRNA | AK017394.1 | - | 3 - 4 | 320 | 110 | |
| AK076926.1 | - | 4 - 5 | 402 | 110 | ||
| AK154463.1 | - | 3 - 4 | 323 | 110 | ||
| BC120528.1 | - | 3 - 4 | 247 | 40129451 | 110 | |
| BC120530.1 | - | 3 - 4 | 247 | 40129453 | 110 | |
| BU053145.1 | - | 3 - 4 | 330 | 110 | ||
| CJ147966.1 | - | 3 - 4 | 350 | 110 | ||
Set Membership: |
|