Search
Search

Mastl| Gene Symbol: Mastl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025979.4 | NP_080255.3 | 10 - 11 | 2502 | 147 | |
| GenBank mRNA | AK006448.1 | - | 5 - 6 | 1603 | 147 | |
| AK012597.1 | - | 5 - 6 | 1603 | 147 | ||
| AK030140.1 | - | 10 - 11 | 2397 | 147 | ||
| AK150580.1 | - | 2 - 3 | 1490 | 147 | ||
| BC066834.1 | - | 1759 | 147 | |||
| BC086483.1 | - | 1771 | 147 | |||
| BC103779.1 | - | 10 - 11 | 2378 | 30480071 | 147 | |
Set Membership: |
|