Search
Search

Mastl| Gene Symbol: Mastl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025979.4 | NP_080255.3 | 4 - 5 | 708 | 129 | |
| GenBank mRNA | AK030140.1 | - | 4 - 5 | 603 | 129 | |
| AK078365.1 | - | 4 - 5 | 582 | 129 | ||
| BC103779.1 | - | 4 - 5 | 584 | 30480071 | 129 | |
| CN536399.1 | - | 4 - 5 | 481 | 129 | ||
Set Membership: |
|