Search
Search

Acbd5| Gene Symbol: Acbd5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102436.1 | NP_001095906.1 | 8 - 9 | 1001 | 76 | |
| NM_001102437.1 | NP_001095907.1 | 8 - 9 | 1128 | 76 | ||
| NM_001102438.1 | NP_001095908.1 | 8 - 9 | 1128 | 76 | ||
| NM_028793.3 | NP_083069.1 | 8 - 9 | 1001 | 76 | ||
| GenBank mRNA | AK005001.2 | - | 7 - 8 | 917 | 76 | |
| AK050450.1 | - | 8 - 9 | 967 | 76 | ||
| AK147839.1 | - | 8 - 9 | 1128 | 76 | ||
| BC035202.1 | - | 8 - 9 | 1057 | 76 | ||
| BC053518.1 | - | 8 - 9 | 1095 | 76 | ||
| BC061484.1 | - | 7 - 8 | 899 | 5355466 | 76 | |
| BC084584.1 | - | 8 - 9 | 933 | 5257773 | 76 | |
| BC090641.1 | - | 8 - 9 | 952 | 76 | ||
Set Membership: |
|