Search
Search

Apbb1ip| Gene Symbol: Apbb1ip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019456.2 | NP_062329.2 | 9 - 10 | 1251 | 150 | |
| GenBank mRNA | AF020313.1 | - | 6 - 7 | 856 | 149 | |
| AK089987.1 | - | 1 - 2 | 125 | 150 | ||
| BC003991.1 | - | 6 - 7 | 902 | 150 | ||
| BC023110.1 | - | 9 - 10 | 1140 | 3707478 | 150 | |
Set Membership: |
|