Search
Search

Lin7c| Gene Symbol: Lin7c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011699.3 | NP_035829.1 | 2 - 3 | 157 | 77 | |
| GenBank mRNA | AF087695.1 | - | 2 - 3 | 157 | 77 | |
| AF173083.1 | - | 2 - 3 | 164 | 77 | ||
| AK015399.1 | - | 2 - 3 | 176 | 77 | ||
| AK078923.1 | - | 2 - 3 | 177 | 77 | ||
| AK083299.1 | - | 2 - 3 | 78 | |||
| AK128989.1 | - | 2 - 3 | 177 | 77 | ||
| AK133531.1 | - | 2 - 3 | 166 | 77 | ||
| AK135810.1 | - | 2 - 3 | 183 | 77 | ||
| AK161076.1 | - | 2 - 3 | 138 | 77 | ||
| BC046966.1 | - | 2 - 3 | 157 | 6493071 | 77 | |
| BC052471.1 | - | 2 - 3 | 167 | 77 | ||
Set Membership: |
|