Search
Search
| Gene Symbol: Itpa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025922.2 | NP_080198.2 | 6 - 7 | 440 | 83 | |
| GenBank mRNA | AB100501.1 | - | 6 - 7 | 452 | 83 | |
| AK008279.1 | - | 6 - 7 | 480 | 83 | ||
| AK009248.1 | - | 6 - 7 | 452 | 83 | ||
| AK138101.1 | - | 6 - 7 | 685 | 83 | ||
| AK153904.1 | - | 6 - 7 | 459 | 83 | ||
| BC026508.1 | - | 6 - 7 | 440 | 4512398 | 83 | |
| BC094466.1 | - | 6 - 7 | 428 | 4936811 | 83 | |
Set Membership: |
|