Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01329684_m1
          See other NLRP12 GE Assays ›
          Gene Symbol:
          Nlrp12
          Gene Name:
          NLR family, pyrin domain containing 12
          Gene Aliases:
          Nalp12, PYPAF7
          Chromosome Location:
          Chr.7: 3218643 - 3250159 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01329684_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 4,430.00
          Your Price
          Online offer:
          4430.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Nlrp12
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001033431.1 NP_001028603.1 5 - 6 2595 84

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 71 and 85 Mus musculus Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          release of cytoplasmic sequestered NF-kappaB
          negative regulation of signal transduction
          negative regulation of protein autophosphorylation
          negative regulation of NF-kappaB transcription factor activity
          dendritic cell migration
          regulation of I-kappaB kinase/NF-kappaB signaling
          negative regulation of I-kappaB kinase/NF-kappaB signaling
          regulation of cysteine-type endopeptidase activity involved in apoptotic process
          positive regulation of MHC class I biosynthetic process
          negative regulation of interleukin-6 biosynthetic process
          negative regulation of Toll signaling pathway
          negative regulation of cytokine secretion
          negative regulation of interleukin-1 secretion
          negative regulation of inflammatory response
          negative regulation of ERK1 and ERK2 cascade
          cellular response to cytokine stimulus
          negative regulation of NIK/NF-kappaB signaling
          nucleotide binding
          protein binding
          ATP binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0700019123.mm.1 NM_001033431.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700019123.mm.1 NM_001033431.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700019123.mm.1 TC0700002124.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700019117.mm.1 NM_001033431.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700019117.mm.1 NM_001033431.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700019117.mm.1 TC0700002124.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700019125.mm.1 NM_001033431.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700019125.mm.1 NM_001033431.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700019125.mm.1 TC0700002124.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700019120.mm.1 NM_001033431.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700019120.mm.1 NM_001033431.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700019120.mm.1 TC0700002124.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700019126.mm.1 NM_001033431.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700019126.mm.1 NM_001033431.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700019126.mm.1 TC0700002124.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700019118.mm.1 NM_001033431.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700019118.mm.1 NM_001033431.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700019118.mm.1 TC0700002124.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700019124.mm.1 NM_001033431.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700019124.mm.1 NM_001033431.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700019124.mm.1 TC0700002124.mm.1 MTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline