| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0200005341.mm.1 |
NM_001289547.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200005341.mm.1 |
NM_001289549.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200005341.mm.1 |
NM_016973.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200005341.mm.1 |
NR_110347.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200005341.mm.1 |
TC0200000587.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200005335.mm.1 |
NM_001025310.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200005335.mm.1 |
NM_001289547.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200005335.mm.1 |
NM_001289548.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200005335.mm.1 |
NM_001289549.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200005335.mm.1 |
NM_016973.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200005335.mm.1 |
NR_110347.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200005335.mm.1 |
TC0200000587.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200005350.mm.1 |
NM_001025310.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200005350.mm.1 |
NM_001025311.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200005350.mm.1 |
NM_001289547.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200005350.mm.1 |
NM_001289548.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200005350.mm.1 |
NM_001289549.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200005350.mm.1 |
NM_016973.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200005350.mm.1 |
NR_110347.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200005350.mm.1 |
TC0200000587.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200005340.mm.1 |
NM_001025310.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200005340.mm.1 |
NM_001289547.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200005340.mm.1 |
NM_001289548.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200005340.mm.1 |
NM_001289549.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200005340.mm.1 |
NM_016973.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200005340.mm.1 |
NR_110347.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200005340.mm.1 |
TC0200000587.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200005349.mm.1 |
NM_001025310.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200005349.mm.1 |
NM_001289547.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200005349.mm.1 |
NM_001289548.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200005349.mm.1 |
NM_001289549.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200005349.mm.1 |
NM_016973.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200005349.mm.1 |
NR_110347.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200005349.mm.1 |
TC0200000587.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200005344.mm.1 |
NM_001289549.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200005344.mm.1 |
TC0200000587.mm.1 |
MTA TC |