Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01330148_s1
          See other OLFR367-PS GE Assays ›
          Gene Symbol:
          Olfr367-ps
          Gene Name:
          olfactory receptor 367, pseudogene
          Gene Aliases:
          EG545417, MOR128-1, ORL1283, OTTMUSG00000012147, Olfr367
          Chromosome Location:
          Chr.2: 37266951 - 37271387 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01330148_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr367-ps
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001081010.2 NP_001074479.2 2 981 117

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Mus musculus Both primers and probe map within a single exon

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2150941 NM_001081010.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 339307 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 697747 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 58339 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2150941 NM_001081010.2 mouse_transcriptome
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21141435 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21141400 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21141406 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21141433 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21141398 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21141430 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21141411 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21141429 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21141426 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21141409 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21141441 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21141404 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21141402 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21141396 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21141427 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21141447 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21141437 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21141436 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21141419 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21141421 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21141425 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21141407 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21141408 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21141416 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21141422 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21141445 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21141437 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21141408 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01470986 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21141414 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21141421 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02097883 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21141382 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02097887 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21141411 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21141409 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21141430 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21141422 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21141413 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21141416 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21141406 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21141418 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01876812 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02097888 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21141419 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21141436 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21141427 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21141429 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21141403 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21141435 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21141386 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21141433 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21141400 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10471781 - -
          Affymetrix Mouse Gene 1.1 ST Array 10471780 - -
          Affymetrix Mouse Clariom S Array PSR0200006060.mm.1 NM_001081010.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200006060.mm.1 NM_001081010.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200006060.mm.1 TC0200000683.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200006058.mm.1 NM_001081010.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200006058.mm.1 NM_001081010.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200006058.mm.1 TC0200000683.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200006062.mm.1 NM_001081010.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200006062.mm.1 NM_001081010.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200006062.mm.1 TC0200000683.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200006061.mm.1 NM_001081010.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200006061.mm.1 NM_001081010.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200006061.mm.1 TC0200000683.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          MouseRef-8 v2 Expression BeadChip ILMN_2835002 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2835002 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline