Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01330351_mH
          See other SNAP23 GE Assays ›
          Gene Symbol:
          Snap23
          Gene Name:
          synaptosomal-associated protein 23
          Gene Aliases:
          23kDa, AA408749, SNAP-23, Sndt, Syndet
          Chromosome Location:
          Chr.2: 120567651 - 120601255 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01330351_mH
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Snap23
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001177792.1 NP_001171263.1 2 - 3 237 89
          NM_001177793.1 NP_001171264.1 2 - 3 192 89
          NM_009222.3 NP_033248.1 2 - 3 237 89
          GenBank mRNA AB000822.1 - 1 - 2 112 89
          AB007444.1 - 2 - 3 169 89
          AK017311.1 - 2 - 3 212 89
          AK019162.1 - 2 - 3 238 89
          BC070456.1 - 2 - 3 142 30606358 89
          U73143.1 - 1 - 2 61 89

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200001794.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Snap23

          Panther Classification:

          Molecular Function -

          SNARE protein

          Gene Ontology Categories:

          Function(s) Process(es)

          histamine secretion by mast cell
          protein complex assembly
          transport
          exocytosis
          protein transport
          synaptic vesicle priming
          vesicle-mediated transport
          regulation of exocytosis
          synaptic vesicle fusion to presynaptic active zone membrane
          membrane fusion
          SNAP receptor activity
          protein binding
          syntaxin binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0200013745.mm.1 NM_001177792.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200013745.mm.1 NM_001177792.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200013745.mm.1 NM_001177793.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200013745.mm.1 NM_001177793.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200013745.mm.1 NM_009222.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200013745.mm.1 NM_009222.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0200013745.mm.1 TC0200001794.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200013743.mm.1 NM_001177792.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200013743.mm.1 NM_001177792.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200013743.mm.1 NM_001177793.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200013743.mm.1 NM_001177793.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200013743.mm.1 NM_009222.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200013743.mm.1 NM_009222.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0200013743.mm.1 TC0200001794.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200013740.mm.1 NM_001177792.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200013740.mm.1 NM_001177792.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200013740.mm.1 NM_009222.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200013740.mm.1 NM_009222.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0200013740.mm.1 TC0200001794.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200013746.mm.1 NM_001177792.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200013746.mm.1 NM_001177792.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200013746.mm.1 NM_001177793.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200013746.mm.1 NM_001177793.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200013746.mm.1 NM_009222.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200013746.mm.1 NM_009222.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0200013746.mm.1 TC0200001794.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200013751.mm.1 NM_001177792.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200013751.mm.1 NM_001177792.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200013751.mm.1 TC0200001794.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 413285 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-hgw72:80/100.66.79.134:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline