Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01331479_m1
          See other 4631405J19RIK GE Assays ›
          Gene Symbol:
          4631405J19Rik
          Gene Name:
          RIKEN cDNA 4631405J19 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.2: 93029348 - 93033445 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01331479_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 4631405J19Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_110333.1 5 - 6 798 71
          GenBank mRNA AK028444.1 - 2510 71
          BC100298.1 - 5 - 6 798 71

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200001446.mm.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Mus musculus Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P00433019 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P130759 AK028444.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P130759 BC100298.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P00433019 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21432095 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21432118 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01273986 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21432084 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21432094 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21432127 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21432115 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21432116 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21432067 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21432091 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01441586 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21432133 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21432073 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01441596 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01441605 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21432105 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21432075 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01424261 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21432114 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21432109 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01441592 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21432107 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21432123 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21432089 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21432092 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21432068 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21432086 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01569628 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21432076 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01365264 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01365270 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21432100 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21432102 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21432122 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21432134 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21432065 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01294532 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21432077 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01294523 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21432106 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21432130 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21432079 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21432087 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21432119 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10474062 - -
          Affymetrix Mouse Gene 1.0 ST Array 10474061 - -
          Affymetrix Mouse Gene 1.0 ST Array 10474059 - -
          Affymetrix Mouse Gene 1.0 ST Array 10474063 - -
          Affymetrix Mouse Gene 1.1 ST Array 10474057 - -
          Affymetrix Mouse Clariom S Array PSR0200011133.mm.1 NR_110333.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200011133.mm.1 TC0200001446.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200011132.mm.1 NR_110333.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200011132.mm.1 TC0200001446.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200011137.mm.1 NR_110333.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200011137.mm.1 TC0200001446.mm.1 MTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline