| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1200016910.mm.1 |
NM_001165894.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200016910.mm.1 |
NM_001165894.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200016910.mm.1 |
NM_009652.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200016910.mm.1 |
NM_009652.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200016910.mm.1 |
TC1200002434.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1200016906.mm.1 |
NM_001165894.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200016906.mm.1 |
NM_001165894.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200016906.mm.1 |
NM_009652.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200016906.mm.1 |
NM_009652.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200016906.mm.1 |
TC1200002434.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1200016907.mm.1 |
NM_001165894.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200016907.mm.1 |
NM_001165894.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200016907.mm.1 |
NM_009652.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200016907.mm.1 |
NM_009652.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200016907.mm.1 |
TC1200002434.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1200016903.mm.1 |
NM_001165894.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200016903.mm.1 |
NM_001165894.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200016903.mm.1 |
NM_009652.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200016903.mm.1 |
NM_009652.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200016903.mm.1 |
TC1200002434.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1200016909.mm.1 |
NM_001165894.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200016909.mm.1 |
NM_001165894.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200016909.mm.1 |
NM_009652.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200016909.mm.1 |
NM_009652.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200016909.mm.1 |
TC1200002434.mm.1 |
MTA TC |