Search
Search

Ero1lb| Gene Symbol: Ero1lb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026184.2 | NP_080460.2 | 3 - 4 | 505 | 77 | |
| GenBank mRNA | AF489856.1 | - | 3 - 4 | 311 | 77 | |
| AK004981.1 | - | 482 | 77 | |||
| AK006886.1 | - | 3 - 4 | 368 | 77 | ||
| AK041951.1 | - | 3 - 4 | 481 | 77 | ||
| AK160664.1 | - | 3 - 4 | 521 | 77 | ||
| BC058721.1 | - | 3 - 4 | 505 | 5705507 | 77 | |
| BC111900.1 | - | 3 - 4 | 475 | 40062137 | 77 | |
| BC112426.1 | - | 3 - 4 | 475 | 40062136 | 77 | |
| BC145888.1 | - | 3 - 4 | 456 | 40131191 | 77 | |
| BC145890.1 | - | 3 - 4 | 456 | 40131193 | 77 | |
Set Membership: |
|