Search
Search

Lgals8| Gene Symbol: Lgals8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199043.1 | NP_001185972.1 | 2 - 3 | 642 | 122 | |
| NM_001291055.1 | NP_001277984.1 | 1 - 2 | 144 | 122 | ||
| NM_018886.5 | NP_061374.1 | 2 - 3 | 456 | 122 | ||
| GenBank mRNA | AF218069.1 | - | 1 - 2 | 158 | 122 | |
| AK004782.2 | - | 2 - 3 | 324 | 122 | ||
| AK029526.1 | - | 3 - 4 | 504 | 122 | ||
| AK029685.1 | - | 2 - 3 | 351 | 122 | ||
| AK083350.1 | - | 2 - 3 | 209 | 122 | ||
| AK145881.1 | - | 2 - 3 | 176 | 122 | ||
| AK146746.1 | - | 2 - 3 | 185 | 122 | ||
| AK148939.1 | - | 2 - 3 | 221 | 122 | ||
| BC040243.1 | - | 2 - 3 | 290 | 2582159 | 122 | |
Set Membership: |
|