Search
Search
| Gene Symbol: Ckb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021273.4 | NP_067248.1 | 1 - 2 | 114 | 72 | |
| GenBank mRNA | AK002467.1 | - | 1 - 2 | 77 | 72 | |
| AK014299.1 | - | 1 - 2 | 57 | 72 | ||
| AK148885.1 | - | 1 - 2 | 74 | 72 | ||
| AK152388.1 | - | 1 - 2 | 74 | 72 | ||
| AK153484.1 | - | 1 - 2 | 74 | 72 | ||
| AK161990.1 | - | 1 - 2 | 74 | 72 | ||
| AK166980.1 | - | 1 - 2 | 77 | 72 | ||
| AK167034.1 | - | 1 - 2 | 77 | 72 | ||
| BC015271.1 | - | 1 - 2 | 44 | 4225384 | 72 | |
| BC106109.1 | - | 1 - 2 | 41 | 6395794 | 72 | |
Set Membership: |
|